missing translation for 'onlineSavingsMsg'
Learn More

Methylmercury(II) hydroxide, 1M in H{2}O, Thermo Scientific Chemicals

As a defoliant and epoxidation catalyst, as denaturing agent

44.60 € - 862.29 €

Chemical Identifiers

CAS 1184-57-2
MDL Number MFCD00013924
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15471037
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
013395.XF
1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15451037
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
013395.AB
10 mL
240.20 €
10mL
Please to purchase this item. Need a web account? Register with us today!
15461037
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
013395.AD
50 mL
862.29 €
50mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Methyl mercury(II) hydroxide is used as defoliant and epoxidation catalyst, as denaturing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1184-57-2
MFCD00013924

Specifications

1184-57-2
Liquid
MFCD00013924
232.63
Methylmercury(II) hydroxide
Colorless
CH3HgOH
UN2024
1M in H2O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H311-H331-H373
Fatal if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P261-P301+P310-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURY COMPOUNDS, LIQUID, N.O.S.

missing translation for 'rtecsNumber' : OW4900000

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.