missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Buffer Solutions: Standard pH
GREENER_CHOICE

Storage and refill solutions for pH electrodes

Brand:  Mettler Toledo™ 51302069

 View more versions of this product

Product Code. 12127800

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Completely miscible
  • Comes with Quality inspection certificate
  • NIST traceable
TRUSTED_SUSTAINABILITY
Specifications

Specifications

∼100°C
Buffer Solution
Quality inspection certificate
4
Clear, Red
pH Buffer
30 x 20 mL Sachets
Completely Miscible
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mettler Toledo™ Buffer Solutions: Standard pH > Buffer; pH 4; Packaging: 30 x 20mL sachets; NIST Traceable

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.