missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ Buffer Solutions: Standard pH

Storage and refill solutions for pH electrodes
31.95 € - 73.80 €
Specifications
Boiling Point | ∼100°C |
---|---|
Type | Buffer Solution |
Filling Solution Type | pH Buffer |
Includes | Quality inspection certificate |
Water Soluble | Completely Miscible |
Product Code | Brand | Color | pH Range | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | pH Range | Packaging | Price | Quantity & Availability | |||||
12699235
![]() |
Mettler Toledo™
51350052 |
Clear | 4.006 | 250 mL Bottle |
36.15 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10643131
![]() |
Mettler Toledo™
51350058 |
Clear | 10.012 | 250 mL Bottle |
31.95 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12723161
![]() |
Mettler Toledo™
51302047 |
Clear, Green | 7 | 30 x 20 mL Sachets |
73.80 €
600mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12127800
![]() |
Mettler Toledo™
51302069 |
Clear, Red | 4 | 30 x 20 mL Sachets | N/A | ||||||
12663516
![]() |
Mettler Toledo™
51302069 |
Clear, Red | 4 | 30 x 20 mL Sachets |
73.75 €
600mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Completely miscible
- Comes with Quality inspection certificate
- NIST traceable
Specifications
∼100°C | |
pH Buffer | |
Completely Miscible |
Buffer Solution | |
Quality inspection certificate |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Mettler Toledo™ Buffer Solutions: Standard pH