Learn More
IBA Lifesciences MHC I-Strep HLA-A*0201; WT1 (RMFPNAPYL)
GREENER_CHOICE
MHC class I molecule fused to a Twin-Strep-tag™ and refolded with a peptide
Brand:  IBA Lifesciences 6-7019-001
Product Code. 15653908
Description
Description
  • MHC I-Strep HLA-A*0201; WT1 (RMFPNAPYL) is a recombinantly expressed MHC class I molecule that presents a peptide from a Wilms tumor antigen 1 (WT1).
  • Our MHC I Streptamer™ approach is based on our Strep-tag technology and facilitates reversible antigen-specific T cell labeling.
  • A Twin-Strep-tag™ fused to the MHC I-Strep permits multimerization of MHC I complexes on a Strep-Tactin™ backbone, resulting in the formation of MHC I Streptamers.
  • For multimerizing MHC I-Streps, Strep-Tactin™ TACS Agarose columns, Strep-Tactin™ Magnetic Microbeads or fluorophore-conjugated Strep-Tactin™ can be used.
  • This allows affinity chromatographic or magnetic isolation or cell staining for flow cytometry analysis and sorting (FACS), depending on the experimental requirements.
  • The addition of biotin reverses the multimerization of MHC class I molecules, leading to their dissociation from the target cells.
  • The resulting label-free cells are highly viable and fully functional.
  • Specifications
    Specifications
    40 μL
    Fluorescent reagent for staining, magnetic T cell labeling and isolation
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents Certificates
    Special Offers
    Special Offers
    Product Content Correction
    Product Title
    IBA Lifesciences MHC I-Strep HLA-A*0201; WT1 (RMFPNAPYL) > Capacity: 40μL