missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MicroAmp™ TriFlex 3 x 32-Well PCR Reaction Plate

For use with Applied Biosystems™ ProFlex™ 3 x 32-well PCR System, (Cat. No. 4484073) and all Applied Biosystems standard 96-well thermal cyclers

32.20 € - 124.00 €

Specifications

Barcode No Barcode
Certifications/Compliance Certified DNA/RNase Free
Product Line MicroAmp
Color Optical
For Use With (Equipment) Thermal Cycler, Genetic Analyzer, DNA Analyzer
View More Specs

Products 2
Product Code Brand No. per Pack Price Quantity & Availability  
Product Code Brand No. per Pack Price Quantity & Availability  
15835066
View Documents
Applied Biosystems™
A32811
20 Plates
124.00 €
Each
In Stock
to see stock.
Add to basket
 
15818561
View Documents
Applied Biosystems™
A32810
5
32.20 €
Pack of 5
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Offers same thin-wall construction for heat transfer from thermal block to sample
  • Validated with Applied BiosystemsTM PCR thermal cyclers for optimal fit and performance
  • Easy-to-tear dual side tabs for simple separation into three, individual 32-well plates
  • Semi-skirted for easy handling and labeling
  • Certified human DNA-, DNase- and RNase-free
  • Standard 250μL working volume, with 300μL maximum well volume when used with adhesive film
  • Alphanumeric labeling on each 32-well plate for easy well identification
  • 32-well adhesive seal (MicroAmpTM 32-Well Clear Adhesive Film) available for efficient plate sealing
Specifications

Specifications

No Barcode
MicroAmp
Thermal Cycler, Genetic Analyzer, DNA Analyzer
Polypropylene
Room Temperature
Certified DNA/RNase Free
Optical
Semi
32, 96
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Applied Biosystems™ MicroAmp™ TriFlex 3 x 32-Well PCR Reaction Plate

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.