missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ Irradiated IPA Resistant LabMarker
BIOPHARMA

A fine tipped permanent marker with alcohol-resistant ink that marks on cold or wet surfaces, plastic bags and disposable lab-ware.

Brand:  Micronova™ PEN-40IR

89.34 EUR valid until 2025-05-02
Use promo code "24151" to get your promotional price.


Product Code. 12857053

  • 113.00 € / Pack of 10
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

With permanent, quick drying black ink that is resistant to IPA

The Micronova Irradiated IPA Resistant LabMarker features permanent, quick drying, alcohol-resistant ink that marks on a range of surfaces and will not smear on reusable glass, metal or porcelain, but can be scrubbed off. Available gamma irradiated for aseptic areas.

  • Fine-tip, black permanent marker
  • Quick drying, alcohol-resistant ink
  • Marks on cold or wet surfaces, plastic bags, and disposable lab-ware
  • Smear-resistant on glass, metal and porcelain, but can be scrubbed off
  • Available gamma irradiated for aseptic areas
Specifications

Specifications

HDPE
5.25 in.
Reusable
Cleanroom Pen
Fine
Black
Not Autoclavable
Cleanroom and Aseptic Environment Documentation and Labeling
Sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number048535-000 Certificate TypeCertificate of Quality Date04/04/2025 Catalog NumberPEN-40IR
Lot Number048536-000 Certificate TypeCertificate of Quality Date04/04/2025 Catalog NumberPEN-40IR
Lot Number048440-000 Certificate TypeCertificate of Quality Date04/04/2025 Catalog NumberPEN-40IR
Lot Number048439-000 Certificate TypeCertificate of Quality Date14/03/2025 Catalog NumberPEN-40IR
Lot Number046734-000 Certificate TypeCertificate of Quality Date04/02/2025 Catalog NumberPEN-40IR
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Micronova™ Irradiated IPA Resistant LabMarker > LabMarker Irradiated Fine Tip Marker, IPA Resistant Ink, Black

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.