missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Microplate Shakers with 4-Place Platform

For life science, immunoassay. shaking ELISA plates, staining cells for flow cytometry,shaking plates in cold room, food and beverage, histamine in cheese, vitamins in milk testing applications

Brand:  Fisherbrand™ 88861024

581.00 EUR valid until 2025-06-30
Use promo code "24069" to get your promotional price.


 View more versions of this product

Product Code. 15504070

  • 830.00 â‚¬ / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Includes: 4 place microplate clamps on aluminum platform

Description

Description

  • Variable shaking speed: 150 to 1000rpm 
  • Quick and easy fitting or any standard-depth microplates 
  • Continuous or timed operation, with automatic switch off 
  • Display will show last used settings, even after power has been turned off.
  • Continue to run by last set speed and the remaining time when the power recovered.
  • Operating temperature 5°C to 40°C allows for use in refrigerators, cold rooms and non-CO2 incubators
  • Holds up to four microplates  
  • Easy-to-use electronic timer ensures accurate count-down and repeatability of time sensitive incubations
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2.5 mm
Speed Accuracy: ±1 rpm
100/240 V
EU, UK, CN Plug
320 x 260 mm
LED
0 min. to 99:59 hr.
5 kg
150 to 1000 rpm
5°C to 40°C
Aluminum Platform
415 x 293 x 143mm
4 place microplate clamps on aluminum platform
Product Suggestions

Product Suggestions

Videos

Videos

  • Fisherbrand Rockers and Rotators August
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      Fisherbrand Rockers and Rotators August.mp4

    • Fisherbrand microplate shaker
      Video Player is loading.
      Current Time 0:00
      Duration 0:00
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 0:00
       
      1x
      • Chapters
      • descriptions off, selected
      • captions off, selected

        Fisherbrand microplate shaker

      SDS
      Documents Certificates
      View accessory products
      Special Offers

      Special Offers

      Product Content Correction

      Your input is important to us. Please complete this form to provide feedback related to the content on this product.

      Product Title
      Fisherbrand™ Microplate Shakers with 4-Place Platform > Plug type: EU, UK, CN

      By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

      Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.