missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Neurite Outgrowth Staining Kit

The Molecular Probes™ Neurite Outgrowth Staining Kit allows for quick and simple measurement of neurite outgrowth and cell viability in the same sample.

Brand:  Invitrogen™ A15001

249.90 EUR valid until 2025-06-30
Use promo code "24074" to get your promotional price.


Product Code. 13494710

  • 357.00 € / 1000 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Green, Orange
Neurite Outgrowth Staining Kit
1000 assays
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2945662 Certificate TypeCertificate of Analysis Date09/04/2024 Catalog NumberA15001
Lot Number2901460 Certificate TypeCertificate of Analysis Date09/04/2024 Catalog NumberA15001.18,A15001,A15001.22,A15001.36
Lot Number2857316 Certificate TypeCertificate of Analysis Date25/01/2024 Catalog NumberA15001
Lot Number2834649 Certificate TypeCertificate of Analysis Date31/10/2023 Catalog NumberA15001
Lot Number2805447 Certificate TypeCertificate of Analysis Date12/10/2023 Catalog NumberA15001
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Neurite Outgrowth Staining Kit > Neurite Outgrowth Staining Kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.