Learn More
Thermo Scientific™ Microscope Slides, 45°
Premium slides are manufactured through a unique proprietary cleaning process intended to provide consistent results and ease interpretation. This process allows these slides to to be used in routine histology and microbiology applications; and specifically in applications by delivering consistently crisp, featherededge blood smears with good morphology.
Brand: Thermo Scientific™ ABAA000001##02E00MNZ10MH#
Description
- Manufactured from Swiss extra-white soda-lime glass
- Reduced iron content for low self-fluorescence
- Greenish shimmer minimized
- Guaranteed optimum, faultless transparency
- Clean, flat and suitable for immediate use

Specifications
76 mm | |
1 mm | |
No |
26 mm | |
Uncoated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.