Learn More
Labconco™ Microwell Plate Holder
Solid aluminum base tightly holds microplates.
Brand: Labconco™ 7369600
This item is not returnable.
View return policy
Description
Microwell Plate Holder- Made of solid aluminum
- Eliminates air gap on bottom of plate for excellent heat transfer to microplate
- Gives temperature stability to small samples that are susceptible to meltback
- Microplate required and not included
Specifications
Microplate Holder | |
FreeZone™ Freeze Dryers | |
5.7 in. | |
1.78 cm | |
10.16 cm | |
1.3 lb. |
Aluminum | |
Solid aluminum block that holds microwell plate. Microwell plate not included. | |
0.7 in. | |
4 in. | |
2 lb. |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Product Title
Labconco™ Microwell Plate Holder > Microwell Plate Holder
Spot an opportunity for improvement?Share a Content Correction