Learn More
Fisherbrand™ Midi Centrifuge
Achieve max speed with minimum space. Fisherbrand™ Midi Centrifuge is ideal for protocols calling for fast spins with a higher RCF requirement. Ideal for micro-filter cell separations, PCR, clinical applications and HPLC protocols. Includes two rotors: standard 12-place rotor with lid and eight-place strip rotor for four 0.2mL tube strips or 32 single tubes.
Brand:  Fisherbrand™ HS120119
783.89 EUR valid until 2025-07-11
Use promo code "25721" to get your promotional price.
Product Code. 12972041
  • 959.00 € / Each
Edge
Explore more special offers
Description
Description
  • 5 Year Warranty
  • Compact footprint and brush-less motor for quiet operation and low vibration
  • Select and set the digital speed display in RCF or RPM
  • Sample imbalance and tilt safety cut-out
  • Safety interlock lid prevents opening while rotor is turning
  • Back lit display showing time, speed and operational information on the same screen
  • Quick spin button for short spin requirements
  • UV resistant plastic, increased useful life under harsh light conditions
  • Push-button lid release
  • Manual override allows user to open lid in case of power failure
Specifications
Specifications
12 x 1.5/2 mL Tubes (Rotor 1), 8 places for 4 x 0.2 mL Tube Strips or 32 x 0.2 mL Single Tubes (Rotor 2)
Digital
Rotors, 12 x 0.2mL and 12 x 0.5mL tube adapters, universal double insulated power adapter with four interchangeable plugs
171 mm
Yes
For PCR, microfilter separation, clinical applications and HPLC protocols.
9,800 x g
Midi Centrifuge
100/240 V 50/60 Hz
500 to 12,500 rpm
114 x 171 x 203 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Midi Centrifuge > Dimensions: 104H x 153W x 128mmD