missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Biosciences™ DAPI Solution (RUO)
Nucleic acid stain that binds to A-T rich regions of DNA along the minor groove.
Brand: BD Biosciences™ 564907
108.36 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Nucleic acid stain that binds to A-T rich regions of DNA along the minor groove.
- DAPI is predominantly impermeant to live cells, allowing it to be used as a viability dye in unfixed cells to discriminate intact from membrane-compromised cells.
- Note, however, that high concentrations of the dye may still enter intact cells.
- Additionally, DAPI may be used to analyze DNA content in fixed cells, or as a nuclear counterstain in imaging or flow cytometry.
- When bound to double-stranded DNA, DAPI has an excitation wavelength maximum of 358 nm and an emission maximum of 461 nm.
- DAPI is also well excited by the violet laser line (eg, 405 nm). Note that DAPI also binds RNA.
- Under these conditions, DAPI emits maximally at 500 nm, but with less intensity than when bound to double-stranded DNA.

Specifications
1.0 mg/mL | |
Flow cytometry, bioimaging, immunofluorescence | |
Avoid multiple freeze-thaws of product. The product should be kept undiluted at -20°C for long term storage, and it may be kept undiluted at 4°C for short term storage. |
Nucleic acid stain | |
1 mg |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BD Biosciences™ DAPI Solution (RUO) > 1 mg
Spot an opportunity for improvement?Share a Content Correction