Learn More
Thermo Scientific™ Molecular BioProducts Snap-Cap Plain Centrifuge Tubes
Brand: Thermo Scientific™ 3439A
Description
- With flat cap
- No frosting or graduations
- Certified RNase/DNase, RNA/DNA, ATP and pyrogen free

Specifications
Flat | |
0.6 mL | |
Polypropylene | |
Bag | |
No |
RNase, DNase, RNA, DNA, ATP, pyrogen-free | |
Assorted | |
Flat cap | |
Sterile | |
Flat Cap |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.