missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ionomycin, Calcium Salt

Ionomycin is an effective Ca2+ ionopohre that is commonly used to both modify intracellular Ca2+ concentrations and to calibrate fluorescent Ca2+ indicators when studying the regulatory properties of Ca2+ in cellular processes.

Brand:  Invitrogen™ I24222

215.08 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.



Product Code. 10134232

  • 283.00 € / 1mg

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Ionomycin is an effective Ca2+ ionopohre that is commonly used to both modify intracellular Ca2+ concentrations and to calibrate fluorescent Ca2+ indicators when studying the regulatory properties of Ca2+ in cellular processes. Ionomycin also transports Pb2+ and some other divalent cations, as well as several lanthanide series trivalent cations, at efficiencies that are greater than or equal to those for Ca2+.

To make a concentrated stock solution, dissolve in either high quality anhydrous DMSO (1.6 mg/mL) or 100% ethanol (20 mg/mL). Store frozen, desiccated and protected from light.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 mg
Ionophore
Solid
747.08
Room Temperature
C41H70CaO9
Calcium Ionophore
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3011773 Certificate TypeCertificate of Analysis Date24/10/2024 Catalog NumberI24222
Lot Number2810932 Certificate TypeCertificate of Analysis Date13/12/2023 Catalog NumberI24222
Lot Number2725286 Certificate TypeCertificate of Analysis Date13/06/2023 Catalog NumberI24222
Lot Number2544410 Certificate TypeCertificate of Analysis Date10/10/2022 Catalog NumberI24222
Lot Number2420640 Certificate TypeCertificate of Analysis Date17/12/2021 Catalog NumberI24222
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Ionomycin, Calcium Salt > 1 mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.