missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Octadecyl Rhodamine B Chloride (R18)

Lipophilic octadecyl rhodamine B binds to membranes with fluorophore at aqueous interface and alkyl tail protruding into lipid interior

Brand:  Invitrogen™ O246

474.60 EUR valid until 2025-06-30
Use promo code "24074" to get your promotional price.


Product Code. 11529176

  • 678.00 € / 10mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Cell Analysis, Cell Structure, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection, Membranes (General) & Lipids, Plasma Membrane

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

10 mg
Cell Membranes & Lipids
C46H67ClN2O3
Cell Labeling Reagent
Room Temperature
Molecular Probes™
Solid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2980671 Certificate TypeCertificate of Analysis Date06/07/2024 Catalog NumberO246
Lot Number2600140 Certificate TypeCertificate of Analysis Date31/05/2023 Catalog NumberO246
Lot Number2528073 Certificate TypeCertificate of Analysis Date06/09/2022 Catalog NumberO246
Lot Number2351939 Certificate TypeCertificate of Analysis Date30/06/2021 Catalog NumberO246
Lot Number2298148 Certificate TypeCertificate of Analysis Date11/03/2021 Catalog NumberO246
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Octadecyl Rhodamine B Chloride (R18) > 10mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.