missing translation for 'onlineSavingsMsg'
Learn More
Morpholin-2-ylmethyl-carbamic acid tert-butyl ester, 97%, Thermo Scientific™ Product Code.: 10238903

Morpholin-2-ylmethyl-carbamic acid tert-butyl ester, 97%, Thermo Scientific™

Product Code. 10238903
1 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10238903

Brand: Thermo Scientific Maybridge MO08372DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 173341-02-1
Molecular Formula C10H20N2O3
Molecular Weight (g/mol) 216.28
MDL Number MFCD06808588,MFCD12545849,MFCD12545850
InChI Key IYHJNCQAADULQE-UHFFFAOYNA-N
Synonym tert-butyl morpholin-2-ylmethyl carbamate, tert-butyl morpholin-2-ylmethylcarbamate, 2-n-boc-aminomethylmorpholine, tert-butyl n-morpholin-2-ylmethyl carbamate, tert-butyl morpholin-2-yl methyl carbamate, morpholin-2-ylmethyl-carbamic acid tert-butyl ester, carbamic acid,n-2-morpholinylmethyl-, 1,1-dimethylethyl ester, carbamic acid, n-2-morpholinylmethyl-, 1,1-dimethylethyl ester, n-boc-morpholine-2-methanamine, 2-n-boc-aminomethyl morpholineShow MoreShow Less
PubChem CID 19081107
SMILES CC(C)(C)OC(=O)NCC1CNCCO1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
IUPAC Name tert-butyl N-(morpholin-2-ylmethyl)carbamate
Formula Weight 216.28
Percent Purity 97%
Quantity 1 g
Chemical Name or Material Morpholin-2-ylmethyl-carbamic acid tert-butyl ester
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Morpholin-2-ylmethyl-carbamic acid tert-butyl ester, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.