Learn More
MP Biomedicals™ Dextri-Maltose Supplement
Maltose and dextrins obtained by enzymatic digestion of corn flour by barley malt.
Brand: MP Biomedicals™ 0296004801
This item is not returnable.
View return policy
Description
- Maltodextrin, a polymer chain composed of D-glucose units linked by α (1,4) bonds
- Used in the development of encapsulation and non-encapsulation vehicles for use in the food industry
- Used as a protein Maillard-induced conjugation agent for improved solubility and foaming activity
- As a sugar-matrix forming agent, maltodextrin is used in the development of orally disintegrating tablets (ODT)
- Maltose and dextrins obtained by enzymatic digestion of corn flour by barley malt
Specifications
Animal Diet | |
Maltodextrin, a Polymer chain composed of D-glucose units linked by a (1, 4) bonds, is used in the development of encapsulation and nanencapsulation vehicles for use in the food industry, Maltodextrin is used as a protein Maillard-induced conjugation agent for improved solubility and foaming activity | |
15°C to 30°C |
Course Powder | |
1 kg |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
MP Biomedicals™ Dextri-Maltose Supplement > Dextri-Maltose Supplement, 1 kg
Spot an opportunity for improvement?Share a Content Correction