missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ FastPrep-24™ 5G Bead Beating Grinder and Lysis System

FastPrep-24™ 5G Bead Beating Grinder and Lysis System provides the highest yield of DNA, RNA, and Proteins. It processes any tissue or cellular sample type with highest quality results. It has intuitive software and user-friendly touch screen.

Brand:  MP Biomedicals™ 116005500

Product Code. 15260488

  • 12570.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Product Overview: The FastPrep-24 5G is a versatile sample disruption instrument designed to quickly and efficiently homogenize, grind, or lyse biological samples. A completely self-contained system, the FastPrep-24 5G eliminates the risk of cross-contamination and time-consuming clean-up associated with manual lysis methods. Samples are added to individual lysing matrix tubes and placed into sample holder adapters within the instrument. The unique, optimized motion of the FastPrep-24 5G shakes the samples in a tridimensional motion resulting in the collision of the sample material with the lysing matrix particles.
  • Intuitive software makes programming the FastPrep-24 5G fast and simple. The programmable touchscreen display allows for the use of pre-determined or custom homogenization speeds and duration times with a finger touch. Increased power features result in faster processing times and complete lysis of even the toughest samples.
  • Service Plan: One-year warranty is included with purchase.
  • Sample Volume: ∼200 μL to 50 mL; up to ∼25 g
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Plant, Tissue, Bacteria, Fungi, Soil, Feces, Insect
1 to 120 sec.
490 mm
23.6 kg
Touch screen
200 μL to 50 mL, up to 25 g
52 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MP Biomedicals™ FastPrep-24™ 5G Bead Beating Grinder and Lysis System > FastPrep-24™ 5G Bead Beating Grinder and Lysis System

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.