Learn More
MP Biomedicals™ Freund's Complete Adjuvant
Brand:  MP Biomedicals™ 0855828
Product Code. 11424225
  • 172.00 € / 50mL
Explore more special offers
Specifications
2°C to 8°C
Paraffin oil 80 to 90%, Arlacel A 10 to 20%, heat-killed Mycobacterium butyricum 0.1 to 1.5%
Freund's Complete Adjuvant
Used to produce water-in-oil emulsions of immunogens, Immunoassays
50 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
MP Biomedicals™ Freund's Complete Adjuvant > Freund's Complete Adjuva Ea