Learn More
MP Biomedicals™ Freund's Incomplete Adjuvant
Used with subsequent boosts to increase antibody titer following initial animal injection with Freund's Complete Adjuvant
Brand:  MP Biomedicals™ 0855829
Product Code. 11434225
  • 75.80 € / 50mL
Explore more special offers
Specifications
2°C to 8°C
Paraffin oil 80 to 90%, Arlacel A 10 to 20%
Freund's Incomplete Adjuvant
Used with subsequent boosts to increase antibody titer following initial animal injection with Freund's complete adjuvant, immunoassays, as boost injections
25 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
MP Biomedicals™ Freund's Incomplete Adjuvant > Freund's Incomplete Adjuvant, 25mL