missing translation for 'onlineSavingsMsg'
Learn More

IKA™ MultiDrive MT150.5 Milling Tube

The MultiDrive MT150.5 Milling Tube is a plastic milling tube with a stainless steel beater, which is ideally suited for grinding dry samples in combination with the MultiDrive Control Mill. Transparent design allows the grinding material to be continuously observed.

Brand:  IKA™ 0020023173

Product Code. 17247499

  • 84.00 € / Pack of 5
Estimated Shipment: 16-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Autoclavable to minimize risk of cross contamination
  • Capable of grinding samples up to 10 mm and 5 Mohs
  • 5 TS1 Spare Seals are included, extending the lifespan of the tube
  • Requires TC1 Protective Housing to operate (sold separately)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Plastic Milling Tube with Stainless Steel Beater
Dry Milling
Stainless Steel, PA, PC, PTFE, Silicone
150 mL
IKA™ MultiDrive Control Mill
5 Tubes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
IKA™ MultiDrive MT150.5 Milling Tube > MT150.5 Milling Tube; Quantity: 5 Tubes

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.