Learn More
Thermo Scientific™ MycoSEQ™ Assay Service
MycoSEQ Assay Service is a fast and cost-effective way to verify the absence of mycoplasma in cell lines.
Brand: Thermo Scientific™ A44316
Description
- A fast and cost-effective way to verify the absence of mycoplasma in your cell lines
- This service leverages the MycoSEQ Mycoplasma Detection Kit (4460623), a real-time PCR assay designed and validated to meet the requirements of European Pharmacopeia Chapter 2.6.7
- The primers were designed using a proprietary bioinformatics pipeline to detect >90 mycoplasma species with high specificity and no cross-reactivity to closely related bacterial species
The MycoSeq Assay Service offers:
- Fast data delivery – turnaround time of four weeks or less
- Convenient sample prep – just send a frozen cell pellet
- High sensitivity – demonstrated by internal and external validations to detect 10 cfu/mL
- High specificity – includes proprietary Discriminatory Positive Control to differentiate true positive from any potential cross-contamination positives
- Exceptional support – a dedicated project manager for frequent project communication
Specifications
1 sample | |
Assay Service | |
Research Use Only | |
All |
3 Weeks | |
Applied Biosystems™, QuantStudio™ | |
Cell Line Developers |