missing translation for 'onlineSavingsMsg'
Learn More
N-Boc-3-(2-furyl)-L-alanine dicyclohexylammonium salt, 98%, Thermo Scientific™ Product Code.: 15487216

N-Boc-3-(2-furyl)-L-alanine dicyclohexylammonium salt, 98%, Thermo Scientific™

Product Code. 15487216
250 mg
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15487216

Brand: Thermo Scientific Alfa Aesar H62258.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 331730-08-6
Molecular Formula C24H40N2O5
Molecular Weight (g/mol) 436.593
MDL Number MFCD01860894
InChI Key QYEOQPFAYJEDFE-FVGYRXGTSA-N
Synonym dicyclohexylamine s-2-tert-butoxycarbonyl amino-3-furan-2-yl propanoate, boc-l-2-furylalanine-dcha, 2s-2-tert-butoxycarbonyl amino-3-furan-2-yl propanoic acid; dicha, dicycloammonium fur-2-yl-l-alanine, n-boc protected, boc-l-2-furylalanine.dcha, boc-beta-2-furyl-ala-ohcha, boc-l-2-furylalanine dicyclohexylamine salt, boc-3-2-furyl-l-alanine dicyclohexylamine, boc-3-2-furyl-l-alaninedicyclohexylaminesalt, boc-beta-2-furyl-ala-oh dicyclohexylammonium saltShow MoreShow Less
PubChem CID 16211161
IUPAC Name N-cyclohexylcyclohexanamine;(2S)-3-(furan-2-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC=CO1)C(=O)O.C1CCC(CC1)NC2CCCCC2

Specifications

Color White
Quantity 250 mg
Formula Weight 436.58
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-Boc-3-(2-furyl)-L-alanine dicyclohexylammonium salt

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number81301347 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Boc-3-(2-furyl)-L-alanine dicyclohexylammonium salt, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.