missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(salicylidene)ethylenediaminecobalt(II), 96%, Thermo Scientific Chemicals

31.16 € - 533.00 €

Chemical Identifiers

CAS 14167-18-1
Molecular Formula C16H14CoN2O2
Molecular Weight (g/mol) 325.23
MDL Number MFCD00000009
InChI Key NPAQSKHBTMUERN-UHFFFAOYSA-L
Synonym Salcomine
PubChem CID 57448789
SMILES [Co++].[O-]C1=CC=CC=C1C=NCCN=CC1=CC=CC=C1[O-]
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11364858
View Documents
Thermo Scientific Alfa Aesar
043174.03
1 g
31.16 €
1g
Please to purchase this item. Need a web account? Register with us today!
11344858
View Documents
Thermo Scientific Alfa Aesar
043174.06
5 g
70.05 €
5g
Please to purchase this item. Need a web account? Register with us today!
11334858
View Documents
Thermo Scientific Alfa Aesar
043174.14
25 g
202.01 €
25g
Please to purchase this item. Need a web account? Register with us today!
11354858
View Documents
Thermo Scientific Alfa Aesar
043174.22
100 g
533.00 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Salcomine catalyzes the oxidation of 2,6-disubstituted phenols by dioxygen. Experimental inhibitor of human cytμloviris proteinase activity. Metal complex dyes used as redox catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14167-18-1
325.23
NPAQSKHBTMUERN-UHFFFAOYSA-L
57448789
C16H14CoN2O2
MFCD00000009
Salcomine
[Co++].[O-]C1=CC=CC=C1C=NCCN=CC1=CC=CC=C1[O-]

Specifications

14167-18-1
Odorless
[-CH2N=CH(C6H4O-)]2Co
Salcomine
NPAQSKHBTMUERN-UHFFFAOYSA-L
cobalt;(6Z)-6-[[2-[[(E)-(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino]ethylamino]methylidene]cyclohexa-2,4-dien-1-one
57448789
96%
N,N'-Bis (salicylidene)ethylenediaminecobalt (II)
Black
C16H14CoN2O2
MFCD00000009
Insoluble in water. Soluble in benzene,chloroform,and pyridine.
[Co++].[O-]C1=CC=CC=C1C=NCCN=CC1=CC=CC=C1[O-]
325.23
325.23
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332-H317
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.

P201-P202-P261-P264b-P270-P271-P272-P280g-P281-P301+P312-P302+P352-P304+P340-P308+P313-P330-P333+P313-P363-P501c

H302+H332-H317-H350

missing translation for 'einecsNumber' : 238-012-5

missing translation for 'rtecsNumber' : GG0590000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.