Learn More
Thermo Scientific™ Nalgene™ Heavy-Duty Rectangular LLDPE Tank with Covers
Versatility at a lower cost than stainless-steel tanks.
156.00 € - 564.00 €
Specifications
Type Tank with Cover
Material LLDPE
View More Specs
Products 8
Product Code Brand Capacity (Metric) Width (Metric) Length (Metric) Height (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Width (Metric) Length (Metric) Height (Metric) Price Quantity & Availability  
10272701
View Documents
Thermo Scientific™
14100-0002
8 L 20 cm 20 cm 20 cm
156.00 €
Each
Estimated Shipment: 18-04-2025
to see stock.
Add to basket
10324501
View Documents
Thermo Scientific™
14100-0005
20 L 25 cm 25 cm 36 cm
319.00 €
Each
In Stock
to see stock.
Add to basket
10779041
View Documents
Thermo Scientific™
14100-0010
30 L 30 cm 30 cm 30 cm
199.00 €
Each
In Stock
to see stock.
Add to basket
10109860
View Documents
Thermo Scientific™
14100-0015
38 L 30 cm 45 cm 30 cm
257.00 €
Each
In Stock
to see stock.
Add to basket
10040671
View Documents
Thermo Scientific™
14100-0020
50 L 30 cm 61 cm 30 cm
336.00 €
Each
In Stock
to see stock.
Add to basket
10080701
View Documents
Thermo Scientific™
14100-0040
60 L 30 cm 45 cm 45 cm
318.00 €
Each
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
10536551
View Documents
Thermo Scientific™
14100-0045
120 L 45 cm 61 cm 45 cm
419.00 €
Each
Estimated Shipment: 23-12-2025
to see stock.
Add to basket
10204451
View Documents
Thermo Scientific™
14100-0065
160 L 45 cm 61 cm 61 cm
564.00 €
Each
Estimated Shipment: 23-12-2025
to see stock.
Add to basket
Description
Description
Versatility at a lower cost than stainless-steel tanks. Impact- and abrasion-resistant Thermo Scientific™ Nalgene™ Heavy-Duty Cylindrical LLDPE Tanks withstand most acid, alcohols and bases. Offered in sizes from 19 to 208L.
Features:
  • Molded as one piece, no seams
  • Nominal wall thickness is 5/32 in. (3.97mm)
  • Stepped flange at top provides reinforcement and grip for lifting
  • Not autoclavable
Warranty:
90 days
Note:
Tanks used at elevated temperatures or with high specific-gravity liquids should be externally supported.
Specifications
Specifications
Tank with Cover
LLDPE
Videos
SDS
Documents
Documents
Product Certifications
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Caution:
Plastic tanks are generally subject to more severe conditions that plastic labware; exposure is constant, stresses are greater and different classes and concentrations of chemicals are involved. Please pay special attention to chemical compatibility.
Product Content Correction
Product Title
Thermo Scientific™ Nalgene™ Heavy-Duty Rectangular LLDPE Tank with Covers