Learn More
Thermo Scientific™ Nalgene™ Stopcocks
Develop in-house fluid transfer systems for accurate filling or dispensing. These stopcocks provide reliable, accurate liquid flow control.
Brand: Thermo Scientific™ 6460-0002
Description
Permit reliable, accurate liquid flow control in liquid handling systems by using Thermo Scientific™ Nalgene™ Stopcocks. They are ideal for use in developing in-house fluid transfer systems for one-way fluid path for accurate filling or dispensing. The two-way stopcocks feature serrated tubulation on each end.
- Serrated tubulation on each end accepts 1/2″ through 5/16″ tubing
- Polypropylene with Teflon™ TFE plug
- Autoclavable
Specifications
Stopcock | |
Polypropylene | |
6 | |
Polypropylene |
2 mm | |
1 | |
In-House Fluid Transfer System |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
7282869010 | Certificate of Compliance | 25/05/2018 | 6460-0002 |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.