Learn More
Thermo Scientific™ NativePac OBE-1 SEC Column for Online Sample Preparation
Perform Native-MS analysis of biomacromolecules with the NativePac OBE-1 SEC Column designed for rapid exchange of non-volatile buffer components with a volatile buffer.
Brand: Thermo Scientific™ 43803-052130
273.38 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
The Thermo Scientific™ NativePac OBE-1 Column is a Size Exclusion Chromatography (SEC) column designed for rapid exchange of non-volatile buffer components with a volatile buffer for Native-MS analysis of biomacromolecules. The small pores of the SEC stationary phase maximize separation of the non-volatile components from the sample of interest while the short column length minimizes analysis times to less than 2 minutes. A narrow column ID enables the use of low flow rates for high sensitivity detection and maximizes up time by minimizing buffer consumption.
The NativePac OBE-1 SEC column for online sample preparation offers the following features:
- OBE column for protein molecular weight and structure screening
- Rapid exchange of non-volatile buffer components with a volatile buffer for Native-MS analysis
- Short column length minimizes analysis times and buffer consumption
- Narrow column ID enables low flow rates for high sensitivity
- Small SEC stationary phase pore size maximizes separation of non-volatile components from samples of interest

Specifications
Analytical Column | |
50 mm | |
Spherical | |
2 to 8 | |
Proprietary Diol | |
Online Buffer Exchange |
2.1 mm | |
1450 psi (100 bar) | |
3 μm | |
80 Å | |
Spherical, Fully Porous, Ultrapure Silica | |
Size Exclusion |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.