missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Neurobasal™-A Medium, minus phenol red

Neurobasal™-A Medium, minus phenol red
Brand: Gibco™ 12349015
This item is not returnable.
View return policy
Description
Neurobasal™-A Medium Minus Phenol Red is a basal media that meets the special requirements of post-natal and adult brain neuronal when used with GIBCO™ B-27™ Supplement, but without Phenol Red. Neurobasal™-A Medium can be used to grow neuronal cells from hippocampus, cortex and other regions of the brain. Neurobasal™-A Medium allows for both long and short term maintenance of homogeneous populations of neuronal cells without the need of an astrocyte feeder layer.Use Neurobasal™-A Medium Minus Phenol Red for applications in which the presence of Phenol Red will interfere with your results.
Add Your Own Supplements
All Neurobasal™ media contain no L-glutamine, L-glutamic acid, or aspartic acid. Neurobasal™ media must be supplemented with 0.5 mM L-glutamine or GlutaMAX™-I Supplement and either a serum-free supplement (such as B-27™ Supplement or N-2 Supplement), or with serum. For initial plating, you should add 25μM glutamic acid.
Quality Control Testing
All Neurobasal™ media are tested for pH, osmolality, endotoxin, and the absence of bacterial and fungal contamination.
We also provide Neurobasal™ -A Medium with Phenol Red.
Neurobasal™-A Medium is optimized for post-natal and adult brain neurons. For growing pre-natal and embryonic neurons, we provide Neurobasal™ Medium.
See the entire Neurobasal™ product line.
Learn more about neural cell culture products from Invitrogen at www.invitrogen.com/neuralculture.
For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.

Specifications
Neural Cells | |
Liquid | |
No Phenol Red | |
Ambient | |
Store in refrigerator (2–8°C) and protect from light. |
Serum-free | |
Neurobasal Medium | |
500 mL | |
Endotoxin, Performance, Sterility, Osmolality, pH | |
Neuronal Cell Culture |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 13
User Guide: Neurobasal Medium and Neurobasal‑A Medium
Version: 2.0
File size: 272.7 kB
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2337333 | Certificate of Analysis | 29/01/2025 | 12349015 |
2423521 | Certificate of Analysis | 29/01/2025 | 12349015 |
2423523 | Certificate of Analysis | 29/01/2025 | 12349015 |
2517689 | Certificate of Analysis | 29/01/2025 | 12349015 |
2517691 | Certificate of Analysis | 29/01/2025 | 12349015 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ Neurobasal™-A Medium, minus phenol red > Quantity: 500 mL
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction