missing translation for 'onlineSavingsMsg'
Learn More
NF 110, Tocris Bioscience™ Product Code.: 18085792

NF 110, Tocris Bioscience™

Product Code. 18085792
10 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 18085792

Brand: Tocris Bioscience 2548/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent P2X3 antagonist

High affinity P2X3 receptor antagonist (Ki values are 36, 82 and 4144 nM for P2X3, P2X1 and P2X2 recombinant receptors respectively). Shows no activity at P2Y1, P2Y2 and P2Y11 receptors (IC50 > 10 μM). Potently inhibits α,β-meATP-evoked desensitizing currents in rat DRG neurons (IC50 = 527 nM). Shows antitumor activity against several tumor types. Also inhibits DNA-binding activity of HMGA2 (IC50 = 0.87 μM).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111150-22-2
Molecular Formula C41H28N6Na4O17S4
Molecular Weight (g/mol) 1096.899
InChI Key AQJHZNCSXLBXMY-UHFFFAOYSA-J
Synonym 4,4',4,4'-carbonylbis imino-5,1,3-benzenetriylbis carbonylimino tetrakisbenzenesulfonic acid tetrasodium salt, benzenesulfonic acid,4,4',4,4'-carbonylbis imino-5,1,3-benzenetriylbis carbonylimino tetrakis-,tetrasodium salt 9ci, tetrasodium 4-3-3,5-bis 4-sulfonatophenyl carbamoyl phenyl carbamoyl amino-5-4-sulfonatophenyl carbamoyl benzamido benzenesulfonateShow MoreShow Less
PubChem CID 16066783
IUPAC Name tetrasodium;4-[[3-[[3,5-bis[(4-sulfonatophenyl)carbamoyl]phenyl]carbamoylamino]-5-[(4-sulfonatophenyl)carbamoyl]benzoyl]amino]benzenesulfonateShow MoreShow Less
SMILES C1=CC(=CC=C1NC(=O)C2=CC(=CC(=C2)NC(=O)NC3=CC(=CC(=C3)C(=O)NC4=CC=C(C=C4)S(=O)(=O)[O-])C(=O)NC5=CC=C(C=C5)S(=O)(=O)[O-])C(=O)NC6=CC=C(C=C6)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+]Show MoreShow Less

Specifications

Quantity 10 mg
Formula Weight 1096.9
Percent Purity >98%
Chemical Name or Material NF 110
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
NF 110, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.