missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitrofurantoin, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
25 g
100 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Quantity | 25 g |
IUPAC Name | 1-[(E)-[(5-nitrofuran-2-yl)methylidene]amino]imidazolidine-2,4-dione |
Formula Weight | 238.16 |
Percent Purity | ≥97.5% |
Physical Form | Powder |
Chemical Name or Material | Nitrofurantoin |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 8
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
345215 | Certificate of Analysis | 29/11/2023 | 229380100,229381000,229380250 |
345184 | Certificate of Analysis | 24/11/2023 | 229380100,229381000,229380250 |
344955 | Certificate of Analysis | 08/11/2022 | 229380100,229381000,229380250 |
344542 | Certificate of Analysis | 28/04/2021 | 229380100,229381000,229380250 |
344408 | Certificate of Analysis | 11/09/2020 | 229380100,229381000,229380250 |
Safety and Handling


Product Identifier
- Nitrofurantoin
- Danger
- Acute toxicity Category 4
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Nitrofurantoin, 98%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction