Learn More
Nonanoyl chloride, 96%, Thermo Scientific Chemicals
45.43 € - 132.00 €
Chemical Identifiers
CAS | 764-85-2 |
---|---|
Molecular Formula | C9H17ClO |
Molecular Weight (g/mol) | 176.684 |
MDL Number | MFCD00000768 |
InChI Key | NTQYXUJLILNTFH-UHFFFAOYSA-N |
Synonym | nonanoic acid chloride, pelargonoyl chloride, pelargonyl chloride, nonanoylchloride, n-nonanoyl chloride, unii-0v0dum0390, nonanoylchlorid, nananoyl chloride, nonanic acid chloride, pelargonic acid chloride |
PubChem CID | 69819 |
IUPAC Name | nonanoyl chloride |
SMILES | CCCCCCCCC(=O)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11493248
|
Thermo Scientific Alfa Aesar
A17671.14 |
25 g |
45.43 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11403258
|
Thermo Scientific Alfa Aesar
A17671.22 |
100 g | N/A | ||||||
Description
Nonanoyl chloride is used for the acylation of amine groups of chitosan to simultaneously improve its material properties and adsorption capacity. It was also used in the synthesis of N-nonanoyl piperonylamide, caged vanilloid, N-(4-hydroxy-3-methoxybenzyl)-N-(2-nitrobenzyl)-nonanoylamide or N-(2-Nitrobenzyl)-N-vanillyl-nonanoylamide, n-benzylnonanamide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsNonanoyl chloride is used for the acylation of amine groups of chitosan to simultaneously improve its material properties and adsorption capacity. It was also used in the synthesis of N-nonanoyl piperonylamide, caged vanilloid, N-(4-hydroxy-3-methoxybenzyl)-N-(2-nitrobenzyl)-nonanoylamide or N-(2-Nitrobenzyl)-N-vanillyl-nonanoylamide, n-benzylnonanamide.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
764-85-2 | |
176.684 | |
NTQYXUJLILNTFH-UHFFFAOYSA-N | |
69819 | |
CCCCCCCCC(=O)Cl |
C9H17ClO | |
MFCD00000768 | |
nonanoic acid chloride, pelargonoyl chloride, pelargonyl chloride, nonanoylchloride, n-nonanoyl chloride, unii-0v0dum0390, nonanoylchlorid, nananoyl chloride, nonanic acid chloride, pelargonic acid chloride | |
nonanoyl chloride |
Specifications
764-85-2 | |
0.94 | |
95°C (203°F) | |
1.438 | |
UN3265 | |
Moisture sensitive | |
Reacts with water. | |
CCCCCCCCC(=O)Cl | |
176.684 | |
176.69 | |
Nonanoyl chloride |
-61°C | |
215°C | |
C9H17ClO | |
MFCD00000768 | |
1362621 | |
nonanoic acid chloride, pelargonoyl chloride, pelargonyl chloride, nonanoylchloride, n-nonanoyl chloride, unii-0v0dum0390, nonanoylchlorid, nananoyl chloride, nonanic acid chloride, pelargonic acid chloride | |
NTQYXUJLILNTFH-UHFFFAOYSA-N | |
nonanoyl chloride | |
69819 | |
97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H302+H312+H332-H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 212-131-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
- Nonanoyl chloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Substances or mixtures which\, in contact with water\, emits flammable gases Category 3
Hazard Statement
- H261-In contact with water releases flammable gases.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- EUH029-Contact with water liberates toxic gas.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
- MIXTURE LIST-Contains : Nonanoyl chloride
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.