missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ELISA Buffer Kit

Concentrated

Brand:  Invitrogen™ CNB0011

277.40 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.


Product Code. 10170523

  • 304.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Coating Buffer A (100mL): 1 bottle, Coating Buffer B (100mL): 1 bottle, Assay Buffer (5x) (200mL): 1 bottle, Wash Buffer (25x) (100mL): 3 bottles, Stabilized Chromogen (25mL): 4 bottles, Stop Solution (100mL): 1 bottle.
This item is not returnable. View return policy

Description

Description

Buffer Kit for Antibody Pairs is for use with Invitrogen Antibody Pairs. Kit provides sufficient materials for ten ELISA plates.

Build Your Own Immunoassay, Cell Analysis, Immunoassays

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

10 plates
ELISA
Wet Ice
Concentrate
Coating Buffer A (100 mL): 1 bottle
Coating Buffer B (100 mL): 1 bottle
Assay Buffer (5x) (200 mL): 1 bottle
Wash Buffer (25x) (100 mL): 3 bottles
Stabilized Chromogen (25 mL): 4 bottles
Stop Solution (100 mL): 1 bottle. Store at 2°C to 8°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number446721-000 Certificate TypeCertificate of Analysis Date21/04/2025 Catalog Number
Lot Number441946-000 Certificate TypeCertificate of Analysis Date21/04/2025 Catalog Number
Lot Number439337-000 Certificate TypeCertificate of Analysis Date03/04/2025 Catalog Number
Lot Number433294-000 Certificate TypeCertificate of Analysis Date04/12/2024 Catalog NumberCNB0011
Lot Number430756-000 Certificate TypeCertificate of Analysis Date12/11/2024 Catalog NumberCNB0011
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ ELISA Buffer Kit > 10 plates

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.