missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WesternBreeze™ Chromogenic Kit, anti-mouse

Detect proteins that have been immobilized on membranes (nitrocellulose or PVDF) following western transfer or bound directly from solution (dot blots)

Brand:  Invitrogen™ WB7103

Product Code. 10348452

  • 475.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Detection is accomplished with a ready-to-use BCIP/NBT substrate for alkaline phosphatase. Non-fading purple precipitates develop at the protein bands on the membrane.

WesternBreeze™ chromogenic offers:

  • Clear background
  • High sensitivity-low picogram levels detectable
  • High specificity
    One simple protocol-no optimization required

Proteins, Expression, Isolation and Analysis, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Western Blot Kit
Mouse
AP (Alkaline Phosphatase) Substrate
Nitrocellulose, PVDF
Mouse
The WesternBreeze™ Chromogenic Kits include blocking solutions, primary antibody diluent, ready-to-use secondary antibody solution (anti-mouse, anti-rabbit or anti-goat), ready-to-use chromogenic substrate, wash solutions, and incubation trays. Each kit contains complete reagents for 20 mini-blots. Store the kits at +4°C. Guaranteed stable for 6 months when properly stored.
Wet Ice
Chromogenic
WesternBreeze™
1 kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3029601 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberWB7103
Lot Number3107156 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberWB7103
Lot Number2461349 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberWB7103
Lot Number2461361 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberWB7103
Lot Number2467860 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberWB7103
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ WesternBreeze™ Chromogenic Kit, anti-mouse > Reactivity: mouse; 1 kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.