Learn More
Macherey-Nagel™ NUCLEODUR™ C18 Gravity HPLC Column, 1.8μm Particle Size
Ideal for ultra-fast HPLC separation of ketones. Macherey-Nagel™ NUCLEODUR™ C18 Gravity HPLC Column, 1.8μm Particle Size are analytical EC columns with high purity silica phase for HPLC. The columns are available in a variety of diameters and lengths.
713.00 € - 979.00 €
Specifications
Column Format Analytical Column
Crosslinking Octadecyl
Particle Shape Spherical
pH 1 to 11
Pore Size 110 Å
View More Specs
Products 29
Product Code Brand Length (Metric) Diameter (Metric) Particle Size Price Quantity & Availability  
Product Code Brand Length (Metric) Diameter (Metric) Particle Size Price Quantity & Availability  
12438357
View Documents
Macherey-Nagel™
760083.20
150 mm 2 mm 3 μm
815.00 €
Each
In Stock
to see stock.
Add to basket
12468347
View Documents
Macherey-Nagel™
760081.20
125 mm 2 mm 3 μm
815.00 €
Each
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
12408357
View Documents
Macherey-Nagel™
760082.20
250 mm 2 mm 3 μm
943.00 €
Each
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
10064703
View Documents
Macherey-Nagel™
760076.20
100 mm 2 mm 1.8 μm
815.00 €
Each
In Stock
to see stock.
Add to basket
12757800
View Documents
Macherey-Nagel™
760075.20
150 mm 2 mm 1.8 μm
815.00 €
Each
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
12468337
View Documents
Macherey-Nagel™
760078.20
30 mm 2 mm 1.8 μm
713.00 €
Each
In Stock
to see stock.
Add to basket
12408347
View Documents
Macherey-Nagel™
760079.20
50 mm 2 mm 1.8 μm
713.00 €
Each
In Stock
to see stock.
Add to basket
12747800
View Documents
Macherey-Nagel™
760071.20
75 mm 2 mm 1.8 μm
713.00 €
Each
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
12478347
View Documents
Macherey-Nagel™
760081.30
125 mm 3 mm 3 μm
826.00 €
Each
In Stock
to see stock.
Add to basket
12448357
View Documents
Macherey-Nagel™
760083.30
150 mm 3 mm 3 μm
826.00 €
Each
In Stock
to see stock.
Add to basket
12418357
View Documents
Macherey-Nagel™
760082.30
250 mm 3 mm 3 μm
938.00 €
Each
In Stock
to see stock.
Add to basket
12478337
View Documents
Macherey-Nagel™
760078.30
30 mm 3 mm 1.8 μm
713.00 €
Each
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
12448347
View Documents
Macherey-Nagel™
760080.30
50 mm 3 mm 3 μm
713.00 €
Each
In Stock
to see stock.
Add to basket
10379207
View Documents
Macherey-Nagel™
760079.30
50 mm 3 mm 1.8 μm
713.00 €
Each
In Stock
to see stock.
Add to basket
10658745
View Documents
Macherey-Nagel™
760076.46
100 mm 4.6 mm 1.8 μm
887.00 €
Each
In Stock
to see stock.
Add to basket
12708722
View Documents
Macherey-Nagel™
760084.46
100 mm 4.6 mm 3 μm
887.00 €
Each
In Stock
to see stock.
Add to basket
12498347
View Documents
Macherey-Nagel™
760081.46
125 mm 4.6 mm 3 μm
887.00 €
Each
In Stock
to see stock.
Add to basket
10538403
View Documents
Macherey-Nagel™
760083.46
150 mm 4.6 mm 3 μm
887.00 €
Each
In Stock
to see stock.
Add to basket
10235962
View Documents
Macherey-Nagel™
760082.46
250 mm 4.6 mm 3 μm
979.00 €
Each
In Stock
to see stock.
Add to basket
12498337
View Documents
Macherey-Nagel™
760078.46
30 mm 4.6 mm 1.8 μm
779.00 €
Each
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
12428347
View Documents
Macherey-Nagel™
760079.46
50 mm 4.6 mm 1.8 μm
779.00 €
Each
In Stock
to see stock.
Add to basket
10180153
View Documents
Macherey-Nagel™
760080.46
50 mm 4.6 mm 3 μm
779.00 €
Each
In Stock
to see stock.
Add to basket
12718722
View Documents
Macherey-Nagel™
760086.46
75 mm 4.6 mm 3 μm
779.00 €
Each
Estimated Shipment: 18-02-2025
to see stock.
Add to basket
12488347
View Documents
Macherey-Nagel™
760081.40
125 mm 4 mm 3 μm
826.00 €
Each
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
12458357
View Documents
Macherey-Nagel™
760083.40
150 mm 4 mm 3 μm
826.00 €
Each
In Stock
to see stock.
Add to basket
12428357
View Documents
Macherey-Nagel™
760082.40
250 mm 4 mm 3 μm
938.00 €
Each
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
12488337
View Documents
Macherey-Nagel™
760078.40
30 mm 4 mm 1.8 μm
713.00 €
Each
In Stock
to see stock.
Add to basket
12418347
View Documents
Macherey-Nagel™
760079.40
50 mm 4 mm 1.8 μm
713.00 €
Each
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
12458347
View Documents
Macherey-Nagel™
760080.40
50 mm 4 mm 3 μm
713.00 €
Each
In Stock
to see stock.
Add to basket
Description
Description
These columns are suitable for overall sophisticated analytical separations, including ultra-fast HPLC separation of ketones. The compound classes separated include: pharmaceuticals (e.g., analgesics, antiinflammatory drugs, antidepressants), herbicides, phytopharmaceuticals, and immunosuppressants.
  • Suitable for LC/MS and HPLC at pH extremes
  • Superior base deactivation
  • High density octadecyl modification
  • Ideal for method development
Specifications
Specifications
Analytical Column
Spherical
110 Å
Acetonitrile/water
L1
Silica
Reversed Phase
Reversed Phase
EC Column
Octadecyl
1 to 11
1 Ea.
340 m²/g
C18 Gravity
18%
Yes
Stainless Steel