missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ 14mL Round-Bottom Tube

Perform critical cellular applications with these 14mL round-bottom tubes.

Brand:  Thermo Scientific™ 150268

177.81 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.


 View more versions of this product

Product Code. 11337704

  • 232.00 € / Pack of 500
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Perform critical cellular applications with Thermo Scientific™ Nunc™ 14mL Round-Bottom Tubes, facilitated by the dual position vent/close cap for optimized gas exchange. For research use only

  • Optimized gas exchange with dual position vent/close cap
  • Encased in breathable packaging
  • Certificates of conformity available for every lot
  • For research use only
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LDPE
6000 x g
Mouse Embryo Assay (1-cell stage MEA) tested. Produced under ISO 13485 with full lot traceability. USP Class VI tested. Non-pyrogenic and sterile (SAL 10-6).
95 mm
10
Sterile
Round
17.5 mm
Round Bottom Tube
Dual Position Vent/Close Cap
Polystyrene
500
14 mL
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Schematics & Diagrams

Specification Sheets

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2051669 Certificate TypeCertificate of Compliance Date02/12/2024 Catalog Number150268
Lot Number5162237691 Certificate TypeCertificate of Compliance Date02/12/2024 Catalog Number150268
Lot Number511-2180004 Certificate TypeCertificate of Compliance Date02/12/2024 Catalog Number150268
Lot Number2097133 Certificate TypeCertificate of Compliance Date02/12/2024 Catalog Number150268
Lot Number2131731 Certificate TypeCertificate of Compliance Date02/12/2024 Catalog Number150268
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nunc™ 14mL Round-Bottom Tube > 14mL Round Bottom Tube

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.