Learn More
Thermo Scientific™ Nunc™ 96-Well Polypropylene Sample Processing & Storage Microplates

Store samples safely with these low-binding 96-well polypropylene plates, which will not allow DNA or proteins to bind and will allow for complete sample recovery.
Brand: Thermo Scientific™ 249947
256.23 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Description
Achieve complete sample recovery with Thermo ScientificTM NuncTM 96-Well Polypropylene MicroWellTM Plates, which are the ideal storage plate as polypropylene has a lower binding capacity so proteins or DNA will not bind. Withstands temperatures from -80° to +121°C. Available in a variety of colors for quick identification of storage plates.
Features:
- Shared-wall design increases the well volume
- Wider wells and rounded bottom improve mixing
- Conical bottom well for optimal sample recovery
- Black and white plates available for fluorescent- and luminescent-based applications
- Low binding polypropylene for homogeneous assays and storage

Specifications
Custom Barcoding Available | |
Standard | |
96-well Polypropylene Sample Processing & Storage Plates | |
Autoclavable | |
Low binding PP | |
Yellow | |
ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers | |
450 μL | |
400 μL | |
Non-treated | |
Yes | |
120 | |
120/Cs. | |
Yes | |
ANSI/ SLAS Standard |
Certified RNase and DNase Free. ANSI Compliant. | |
Without Lid | |
8 x 12 | |
20 pk., 120 cs. | |
Nunc | |
96-well plate | |
Polypropylene | |
96 | |
Non-sterile | |
Round, Conical Bottom (V-well) | |
Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation | |
20 | |
For Research Use Only. Not for use in diagnostic procedures. | |
Certified Quality |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1424132 | Certificate of Compliance | 04/03/2025 | 249947 |
1333231 | Certificate of Compliance | 11/12/2024 | 249947 |
1341570 | Certificate of Compliance | 11/12/2024 | 249947 |
1416907 | Certificate of Compliance | 14/11/2024 | 249947 |
1414117 | Certificate of Compliance | 20/09/2024 | 249947 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.