Learn More
Thermo Scientific™ Nunc™ Serological Pipettes
Choose size and packaging options to advance your research with complete confidence. Did you know we now offer NEW Serological Pipettes that offer higher quality and cost savings?
61.90 € - 235.00 €
Product Code | Brand | Volume (Metric) | Packaging Type | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Packaging Type | Price | Quantity & Availability | ||||||
13450287
|
Thermo Scientific™
170353 |
1 mL | Individual Paper/Plastic Wrap | N/A | N/A | N/A | |||||
13470287
|
Thermo Scientific™
170354 |
2 mL | Individual Paper/Plastic Wrap | N/A | N/A | N/A | |||||
13430287
|
Thermo Scientific™
170355 |
5 mL | Individual Paper/Plastic Wrap |
78.40 €
Pack of 200 |
In Stock
Log in to see stock. ![]() |
||||||
13440287
|
Thermo Scientific™
170356 |
10 mL | Individual Paper Wrap |
101.00 €
Pack of 200 |
In Stock
Log in to see stock. ![]() |
||||||
13424776
|
Thermo Scientific™
170357 |
25 mL | Individual Paper/Plastic Wrap |
160.00 €
Pack of 200 |
In Stock
Log in to see stock. ![]() |
||||||
13420297
|
Thermo Scientific™
170358 |
50 mL | Individual Paper/Plastic Wrap | N/A | N/A | N/A | |||||
13480287
|
Thermo Scientific™
170364 |
1 mL | Individual Plastic Wrap |
171.00 €
Pack of 1000 |
In Stock
Log in to see stock. |
||||||
13490287
|
Thermo Scientific™
170365 |
2 mL | Individual Plastic Wrap | N/A | N/A | N/A | |||||
13400297
|
Thermo Scientific™
170366 |
5 mL | Individual Plastic Wrap | N/A | N/A | N/A | |||||
13410297
|
Thermo Scientific™
170367 |
10 mL | Individual Plastic Wrap |
92.00 €
Pack of 200 |
In Stock
Log in to see stock. ![]() |
||||||
13440297
|
Thermo Scientific™
170369 |
50 mL | Individual Plastic Wrap |
176.00 €
Pack of 100 |
In Stock
Log in to see stock. |
||||||
13450297
|
Thermo Scientific™
170371 |
1 mL | Bulk packed, 50/bag | N/A | N/A | N/A | |||||
13460297
|
Thermo Scientific™
170372 |
2 mL | Bulk packed, 50/bag | N/A | N/A | N/A | |||||
13470297
|
Thermo Scientific™
170373 |
5 mL | Bulk packed, 25/bag |
137.00 €
Pack of 500 |
In Stock
Log in to see stock. |
||||||
13480297
|
Thermo Scientific™
170374 |
10 mL | Bulk packed, 25/bag | N/A | N/A | N/A | |||||
13490297
|
Thermo Scientific™
170375 |
25 mL | Bulk packed, 25/bag |
126.00 €
Pack of 200 |
In Stock
Log in to see stock. |
||||||
13400307
|
Thermo Scientific™
170376 |
50 mL | Bulk packed, 25/bag | N/A | N/A | N/A | |||||
15101539
|
Thermo Scientific™
170360 |
5 mL | Shortie Pipet, Individual Paper/Plastic Wrap | N/A | N/A | N/A | |||||
15191529
|
Thermo Scientific™
170361 |
10 mL | Individual Paper/Plastic Wrap | N/A | N/A | N/A | |||||
Description
Select size and packaging options to advance your research with complete confidence when using Thermo Scientific™ Nunc™ Serological Pipettes. These accurate, disposable plastic pipettes are plugged and sterilized.
Features:- 100% virgin polystyrene for maximum clarity
- Certified non-pyrogenic
- Non-cytotoxic
- Sterilized pipettes are Gamma irradiated at a validated dose to ensure Sterility Assurance Level of 10-6
- Bold clear graduations with easy-to-read scale markings
- Non-dripping tip for pipette precision
- Color-coded packaging for ease in sorting and selecting the correct size
- Individually wrapped; paper/plastic or plastic film; peel or pop through for easy opening
- Bulk packed for frequent use; reduces packaging waste
- Shortie pipette with ergonomically friendly design for use under laminar hood
Warranty: 90 days
Specifications
Yes | |
Polystyrene | |
Sterile |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.