Learn More
MP Biomedicals™ NZ Bovine Thombin, Low Specific Activity
Manufactured in a state-of-the-art facility in New Zealand using a chromatographic extraction process, under Quality Systems that are ISO Certified and audited to cGMP Principles.
Brand: MP Biomedicals™ 0219990790
Description
- Thrombin is used throughout the diagnostics industry in a variety of coagulation assays, clotting factor tests, and defibrination of blood or plasma for serum controls
- Also used for site specific cleavage of recombinant fusion proteins, or in biochemical and medical research applications
- Manufactured in New Zealand from bovine plasma of New Zealand origin using prothrombin activated with thromboplastin extracted from bovine lung tissue
- Lyophilized under vacuum in glass vials, or in closed trays as a bulk powder (depending on customer requirements)
- Specific Activity: 90-300 NIH units/mg protein
- Total Protein: > 50%

Specifications
Coagulation Assays, Clotting Factor Tests, Defibrination of Blood/Plasma for Serum Controls, Site Specific Cleavage of Recombinant Fusion Proteins, Biochemical Research, Medical Research | |
Bovine Plasma | |
Bovine | |
Cold Pack | |
Manufactured in a state-of-the-art facility in New Zealand using a chromatographic extraction process, under Quality Systems that are ISO Certified and audited to cGMP Principles. |
500 kU | |
Specific Activity: 90 to 300 units/mg protein | |
Frozen | |
Lyophilized |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- Bovine Thrombin, Low Specific Activity, Diagnostic Use
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Thrombin
Your input is important to us. Please complete this form to provide feedback related to the content on this product.