missing translation for 'onlineSavingsMsg'
Learn More

Oleylamine, approximate C18-content 80-90%, Thermo Scientific Chemicals

36.38 € - 564.00 €

Chemical Identifiers

CAS 112-90-3
Molecular Formula C18H37N
Molecular Weight (g/mol) 267.50
MDL Number MFCD00066507
InChI Key QGLWBTPVKHMVHM-KTKRTIGZSA-N
Synonym 9-octadecenylamine, 9-octadecen-1-amine, cis-9-octadecenylamine, e-octadec-9-en-1-amine, 1-amino-9-octadecene, 9e octadec-9-enylamine, 9-octadecenylamine 8ci, 36505-83-6 hydrofluoride, 3811-68-5 unspecified acetate
PubChem CID 6258392
SMILES CCCCCCCC\C=C/CCCCCCCCN
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10316630
View Documents
Thermo Scientific Acros
129540050
5 mL
36.38 €
5mL
Please to purchase this item. Need a web account? Register with us today!
10400380
View Documents
Thermo Scientific Acros
129541000
100 mL
44.22 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10356580
View Documents
Thermo Scientific Acros
129540010
1 L
219.09 €
1 L
Please to purchase this item. Need a web account? Register with us today!
15254624
View Documents
Thermo Scientific Acros
129540051
5 L
564.00 €
5 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

112-90-3
267.50
QGLWBTPVKHMVHM-KTKRTIGZSA-N
6258392
C18H37N
MFCD00066507
9-octadecenylamine, 9-octadecen-1-amine, cis-9-octadecenylamine, e-octadec-9-en-1-amine, 1-amino-9-octadecene, 9e octadec-9-enylamine, 9-octadecenylamine 8ci, 36505-83-6 hydrofluoride, 3811-68-5 unspecified acetate
CCCCCCCC\C=C/CCCCCCCCN

Specifications

112-90-3
0.8100g/mL
Glass bottle
CH3(CH2)7CH=CH(CH2)8NH2
MFCD00066507
QGLWBTPVKHMVHM-KTKRTIGZSA-N
(E)-octadec-9-en-1-amine
6258392
≥96.0% (primary amine)
Oleylamine
Yellow
Authentic
C18H37N
1.4585 to 1.4625 (20°C, 589nm)
9-octadecenylamine, 9-octadecen-1-amine, cis-9-octadecenylamine, e-octadec-9-en-1-amine, 1-amino-9-octadecene, 9e octadec-9-enylamine, 9-octadecenylamine 8ci, 36505-83-6 hydrofluoride, 3811-68-5 unspecified acetate
CCCCCCCC\C=C/CCCCCCCCN
267.50
267.5
Liquid After Melting
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.