missing translation for 'onlineSavingsMsg'
Learn More

Diba Omnifit™ Q-Series Bottle Caps

Easy operation ON/OFF valves offer a simple way to connect tubing

Encompass_Preferred

POA - 459.53 €

Specifications

Color Blue, White
No. per Case 1/Cs.
Sterility Non-sterile
Autoclavable Not Autoclavable
Disposable Reusable
View More Specs

Requires: Use OF9VF check valve and filter assembly to prevent harmful vapors from escaping and to stop contamination from entering the bottle.


Products 9
Product Code Brand Thread Size (Metric) Includes Price Quantity & Availability  
Product Code Brand Thread Size (Metric) Includes Price Quantity & Availability  
12996291
Encompass_Preferred
View Documents
Diba
00932Q-3V
GL 32 Three valved port with PCTFE rotor N/A N/A N/A View alternative products
Save Up to 
»
15284569
Encompass_Preferred
View Documents
Diba
00938Q-2
Two port
POA
Each
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.

Request a Price for this item here.

15294569
Encompass_Preferred
View Documents
Diba
00938Q-3
Three port
POA
Each
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.

Request a Price for this item here.

12906301
Encompass_Preferred
View Documents
Diba
00945Q-2
GL 45 Two port N/A N/A N/A View alternative products
Save Up to 
»
15274579
Encompass_Preferred
View Documents
Diba
00945Q-2V
GL 45 Two valved port with PCTFE rotor
POA
Each
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.

Request a Price for this item here.

12916301
Encompass_Preferred
View Documents
Diba
00945Q-3
GL 45 Three port N/A N/A N/A View alternative products
Save Up to 
»
12926301
Encompass_Preferred
View Documents
Diba
00945Q-3V
GL 45 Three valved port with PCTFE rotor N/A N/A N/A View alternative products
Save Up to 
»
12936301
Encompass_Preferred
View Documents
Diba
00945Q-4
GL 45 Four port
POA
Each
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.

Request a Price for this item here.

14844601
Encompass_Preferred
View Documents
Diba
00945Q-4V
GL 45 Four valved port with PCTFE rotor
459.53 €
Each
View alternative products
Save Up to 
»
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
 
Description

Description

  • 1/4 in.-28 ports with a stainless-steel thread insert to enable repeated connect/disconnect
  • Easily control the flow of liquid into and out of the bottle with integrated valves
  • Each fluid line can be switched on or off independently
  • Anti-twist design ensures no kinked tubing during cap removal
  • With polypropylene screw cap, chemically inert solid PTFE body, 1/8 O.D. x 0.06 in. I.D. tubing

Order Info

Q Series Bottle Caps with valved ports include PCTFE rotors to permit manual shutoff.

Specifications

Specifications

Blue, White
Non-sterile
Reusable
Q series solvent bottle
1/Cs.
Not Autoclavable
Solvent Waste Cap
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Diba Omnifit™ Q-Series Bottle Caps

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.