Learn More
Hirschmann™ opus™ Titration
Guarantees excellent accuracy during serial dispensing. Dispenser is for motor-driven dispensing directly from the reagent bottle.
Brand: Hirschmann™ 9582010
Description
- Electronic control and titration of volumes
- A light protection window for the valve block is also included in the scope of delivery
- TFT touchscreen with user-friendly menu
- Method memory for nine different titration procedures with storage of all settings
- Media recirculating system for simple and rapid venting without reagent loss
- Free rotation
- Superior materials and reliable piston sealing, even in the case of problematic media
- USB and RS232 interface for PC connection
- Integration in automated procedures possible
- Conformity certified
- Available in three different volumes
Specifications
0.6% | |
opus™ titration, motorized bottle top titration with reagent recirculation system | |
TFT Touchscreen | |
EU, US, UK plug type; 3 thread adapters (A32, A38, S40) | |
0.2% | |
240 V |
Electronic Bottletop Titration | |
10 mL | |
Bottle | |
0.01 mL | |
1 Ea. |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.