Learn More
Eppendorf™ Varispenser™ 2x Bottle-Top Dispenser with Recirculation Valve
Ideal for dispensing aliquots of liquid from large supply bottles.
Brand:  Eppendorf™ 4967000057
556.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15781587
  • 563.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Ideal for dispensing aliquots of liquid from large supply bottles.
  • Recirculation valve
  • Standard GL 45 thread on all sizes for most common bottle threads
  • Safety valve balls in discharge valve prevents leakage when discharge tube is not mounted
  • Screwable air vent cap for easy assembly of drying tube or microfilter
  • Oval-shaped dispenser housing for easy transportation
  • PFA seal of the sliding piston prevents seizing up
  • Fixation slide on interior toothed track for rapid and accurate volume adjustment
  • High chemical resistance of all dispensing components
  • Telescopic aspirating tubes for variable adjustment to bottle height
  • Fully autoclavable without disassembly
  • User adjustment option
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Analog Bottletop Dispenser with Adapters
1
5.0 to 50 mL
Recirculation valve, adapters GL 32, GL 38, S 40, and telescopic intake tube (170 – 330 mm)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Eppendorf™ Varispenser™ 2x Bottle-Top Dispenser with Recirculation Valve > 5-50 mL