Learn More
Thermo Scientific™ Orion™ Metallic Combination Electrodes
Attain stable and drift-free ORP measurements in a variety of sample type.
Brand:  Thermo Scientific™ 977900
 View more versions of this product
Product Code. 10758011
Description
Description
Thermo Scientific™ Orion™ ORP electrodes are available in different body materials to allow for better sample compatibility. The 9678BNWP is a Sure-Flow electrode built with a unique free-flowing liquid-to-liquid junction, that simplifies measurements of dirty or viscous samples by helping to prevent clogging, making them easy to clean. They have rugged epoxy bodies and are best suited for use in strong acids/bases, pH > 12 or pH < 2, food & cosmetic emulsions, pure water and slurries. The 9778BNWP is available in a chemically resistant glass body and best suited for use in wastewater, metal plating and organic solvent samples.
Warranty: 12 months from date of purchase by Buyer or 18 months from date of shipment from Seller, whichever is earlier.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Model 977900; Double platinum KF glass electrode
Dual Platinum
Double Platinum KF Electrode, Glass
Orion Meters
Double
0°C to 40°C
KF coulometric titrations
32°F to 104°F
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Orion™ Metallic Combination Electrodes > Model 977900; Double platinum KF glass electrode