missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ pH Buffer Bottles

Perform accurate, reproducible pH calibrations with these color-coded buffer bottles.

Brand:  Thermo Scientific™ 910104

19.07 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.


 View more versions of this product

Product Code. 10759161

  • 21.11 € / 475mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Use Thermo Scientific™ Orion™ pH buffer bottles to perform accurate, reproducible pH calibrations on your pH meter and pH electrode measurement system. Orion pH buffers are available in 475mL bottles for everyday use and laboratory and bench pH analysis and available in 60 mL bottle 5 packs for occasional use and portable and field pH analysis. Using high-quality pH buffers to calibrate your pH system is the best way to ensure your pH measurements are correct and repeatable.

All Orion pH Buffer Bottles have pH/temperature tables printed on the labels for accurate calibrations and NIST traceability. Manufactured under ISO 9000 quality standards.

Buffer Color Codes:

  • pH 4.01 buffer is pink
  • pH 5.00 buffer is orange
  • pH 7.00 buffer is yellow
  • pH 10.01 buffer is blue

The buffers are available in pint (475mL) bottles, portable five packs of 2 oz. (5 x 60mL) bottles, convenient All-in-One buffer kits, 5 gallon (19L) Cubitainers™ and single-use pouches.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

pH Buffer, bottles
Orion pH 4.01 buffer, color-coded pink, 475mL bottle
pH Meter and Electrode
Liquid
pH Buffer
Bottle
Pink
Accurate pH calibrations and NIST traceability, laboratory and bench analysis
4.01 ± 0.01 pH
475 mL
Water, Potassium Hydroxide, Amaranth Red No. 27, Potassium Hydrogen Phthalate
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberSDZ7 Certificate TypeCertificate of Analysis Date20/01/2025 Catalog Number910104
Lot NumberSDZ6 Certificate TypeCertificate of Analysis Date20/01/2025 Catalog Number910104
Lot NumberSDZ8 Certificate TypeCertificate of Analysis Date20/01/2025 Catalog Number910104
Lot NumberSDZ5 Certificate TypeCertificate of Analysis Date20/01/2025 Catalog Number910104
Lot NumberSDZ2 Certificate TypeCertificate of Analysis Date20/01/2025 Catalog Number910104
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Orion™ pH Buffer Bottles > pH 4.01 Buffer, Color Coded Red, 475mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.