Learn More
Thermo Scientific™ Orion™ Powder Chemistry Packs
Achieve easier and cleaner measurements with pre-measured and ready-to-use powder chemistry packs.
Brand: Thermo Scientific™ AC4P12
Description
Use Thermo Scientific™ Orion™ AQUAfast Powder Pack Reagents with Orion AQUAfast colorimeters and Thermo Scientific Orion AquaMate Vis and UV-Vis spectrophotometers. Powder pack chemistries dissolve quickly in solution, are easy to mix and do not require a stir rod or crushing. Each powder pack is notched for simplified pouch opening in the lab or field. Powder pack reagents measure aluminum, ammonia, chlorine, copper, cyanide, DEHA, iron, manganese, molybdate, nitrite, phosphate, silica and sulfate.
- Equivalent to other widely used and recognized chemistries
- Compatible with Orion™ AQUAfast™ Colorimeters
Specifications
AQUAfast ammonia reagent powder packs, 100 tests | |
Wastewater: Standard Methods 4500-NH3 F-1997 | |
Set of 2: 200 Ammonia Salicylate Powder Packs, 200 Ammonia Cyanurate Powder Packs | |
100 tests | |
AQUAfast 24 mm round vials (Cat. No. AC2V24) |
Ammonia | |
Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast AQ3700 Colorimeter | |
Powder Pack | |
0.01 to 0.8 mg/L N | |
Salicylate |
Safety and Handling
- Ammonia Cyanurate Reagent F10
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : lithium hydroxide monohydrate, sodium dichloroisocyanurate,dihydrate
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.