missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ Gel-Drying Supplies

Use the frames and pre-cut cellophane that come in Thermo Scientific™ Owl™ Gel-Drying Supplies with Owl™ Gel-Drying Kits.

Brand:  Thermo Scientific™ CM-20

 View more versions of this product

Product Code. 11708174

  • 39.35 € / Pack of 50
Estimated Shipment: 15-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Combine the frames and pre-cut cellophane that come in Thermo Scientific™ Owl™ Gel-Drying Supplies with Owl™ Gel-Drying Kits.

  • Frames and pre-cut cellophane available in large (24 × 24cm) or small (14 × 14cm) format
TRUSTED_SUSTAINABILITY
Specifications

Specifications

24 x 24 cm
Cellophane, Precut
24 cm
24 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Owl™ Gel-Drying Supplies > Precut Cellophane; 24 x 24cm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.