Learn More
Oxalyl chloride, 98%, Thermo Scientific Chemicals
Description
Ammonium hexafluorotitanate is used as an anti-corrosive cleaning agent. It is also used for the production of ceramics and glass. Further, it is used in the preparation of synthetic gems.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Oxalyl chloride is mainly used as a catalyst in the oxidation of alcohols to the corresponding aldehydes and ketones. It is actively used for the synthesis of acid chlorides from acids. In the Fridel-Crafts acylation reaction, it reacts with aromatic compounds to get the corresponding acyl choride using aluminum chloride as catalyst. Further, it is utilized to prepare dioxane tetrketone, an oxide of carbon.
Solubility
Oxalyl chloride Slightly miscible with water.Miscible with ether, benzene and chloroform.
Notes
Moisture sensitive. Incompatible with bases, alcohols, water, amines and metals.

Specifications
Specifications
Melting Point | -12°C |
Density | 1.455 |
Boiling Point | 63°C to 64°C |
Assay Percent Range | 0.98 |
Odor | Acrid |
Refractive Index | 1.429 |
Quantity | 1000 g |
UN Number | UN2927 |
Beilstein | 1361988 |
Sensitivity | Moisture sensitive |
Show More |
Safety and Handling



- Oxalyl chloride
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Substance which\, in contact with water\, emits flammable gases Category 1
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H301-Toxic if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- EUH014-Reacts violently with water.
- EUH029-Contact with water liberates toxic gas.
- EUH071-Corrosive to the respiratory tract.
- P231-Handle under inert gas.
- P232-Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P335+P334-Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.
- MIXTURE LIST-Contains : Ethanedioyl dichloride
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.