Learn More
Thermo Scientific™ PageRuler™ Unstained High Range Protein Ladder
A mixture of 8 unstained proteins (60 to 250kDa) for use as size standards in protein electrophoresis (SDS-PAGE) and Western blotting.
Brand: Thermo Scientific™ 26637
136.80 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
The Thermo Scientific™ PageRuler Unstained High Range Protein Ladder is a mixture of 8 unstained proteins (60 to 250kDa) for use as size standards in protein electrophoresis (SDS-PAGE) and Western blotting.
The protein MW markers in this ladder resolve into clearly identifiable, sharp bands when analyzed by SDS-PAGE and stained with coomassie dye or silver. The 150kDa band is of greater intensity and serves as a reference band. Proteins can be detected on Western blots by staining with Ponceau S, coomassie blue or other protein stains. In addition, the ladder proteins contain an integral Step-tag™ II Sequence and may be detected on Western blots using Strep-Tactin™ Conjugates. The PageRuler Unstained High Range Protein Ladder is supplied ready-to-use: no heating, dilution or addition of a reducing agent is required before use.
Highlights:
- Size range 8 proteins spanning 60 to 250kDa
- Ready-to-use supplied in a loading buffer for direct loading on gels
- Sharp bands excellent accuracy for detection with coomassie or silver stains
- Reference band 150kDa band has greater intensity for easy orientation
- Tagged each protein in the ladder contains an integral Strep-tag™ II Sequence
Quality tested each lot evaluated by SDS-PAGE and Western blotting
Includes:
- Proteins (0.02 to 0.05mg/mL of each) in 62.5mM Tris-H3PO4 (pH 7.5 at 25°C), 1mM EDTA, 2% SDS, 100mM DTT, 1mM NaN3, 0.01% bromophenol blue and 33% glycerol
Recommended for:
- Accurate protein sizing on SDS-polyacrylamide gels and Western blots

Specifications
Upon receipt store at -20°C. Product is shipped with an ice pack. | |
Protein Ladder | |
Tris-acetate gels | |
Unstained | |
250, 200, 150, 120, 100, 85, 70, 60 kDa | |
2 x 250 μL | |
Western Blotting, SDS-PAGE |
8 | |
Yes | |
60 to 250 kDa | |
Bolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels | |
PageRuler™ | |
Approved for shipment on Wet or Dry Ice |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2821205 | Certificate of Analysis | 29/01/2025 | 26637_LITL18 PROMO,26637 |
2830870 | Certificate of Analysis | 29/01/2025 | 26637_LITL18 PROMO,26637 |
2831963 | Certificate of Analysis | 29/01/2025 | 26637_LITL18 PROMO,26637 |
2793484 | Certificate of Analysis | 29/01/2025 | 26637_LITL18 PROMO,26637 |
2836263 | Certificate of Analysis | 29/01/2025 | 26637_LITL18 PROMO,26637 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.