Learn More
Palladium(II) acetate, 47.5% Pd, Thermo Scientific Chemicals
149.00 € - 5571.00 €
Chemical Identifiers
CAS | 3375-31-3 |
---|---|
Molecular Formula | C4H6O4Pd |
Molecular Weight (g/mol) | 224.51 |
MDL Number | MFCD00012453 |
InChI Key | YJVFFLUZDVXJQI-UHFFFAOYSA-L |
Synonym | palladium ii acetate, palladium diacetate, palladium acetate, diacetoxypalladium, bis acetato palladium, bisacetylpalladium, diacetatopalladium, palladium 2+ acetate, palladous acetate |
PubChem CID | 167845 |
SMILES | [Pd++].CC([O-])=O.CC([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10767234
|
Thermo Scientific Acros
195180010 |
1 g | Glass bottle |
149.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10522221
|
Thermo Scientific Acros
195180020 |
2 g | Glass bottle |
299.00 €
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10790701
|
Thermo Scientific Acros
195180100 |
10 g | Glass bottle |
1414.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11984351
|
Thermo Scientific Acros
195180500 |
50 g | Glass Bottle |
5571.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3375-31-3 | |
224.51 | |
YJVFFLUZDVXJQI-UHFFFAOYSA-L | |
167845 |
C4H6O4Pd | |
MFCD00012453 | |
palladium ii acetate, palladium diacetate, palladium acetate, diacetoxypalladium, bis acetato palladium, bisacetylpalladium, diacetatopalladium, palladium 2+ acetate, palladous acetate | |
[Pd++].CC([O-])=O.CC([O-])=O |
Specifications
205.0°C | |
C4H6O4Pd | |
MFCD00012453 | |
palladium ii acetate, palladium diacetate, palladium acetate, diacetoxypalladium, bis acetato palladium, bisacetylpalladium, diacetatopalladium, palladium 2+ acetate, palladous acetate | |
YJVFFLUZDVXJQI-UHFFFAOYSA-L | |
palladium(2+) diacetate | |
167845 | |
47.5% (Pd) |
3375-31-3 | |
(CH3CO2)2Pd | |
15,7092 | |
Solubility in water: insoluble. Other solubilities: soluble in acetone, benzene, methylene dichloride,, chloroform, acetonitrile and diethylether, insoluble in alcohols | |
[Pd++].CC([O-])=O.CC([O-])=O | |
224.51 | |
224.51 | |
Palladium(II) acetate, 47.5% Pd |
Safety and Handling
GHS H Statement
Causes serious eye damage.
May cause long lasting harmful effects to aquatic life.
GHS P Statement
Wear eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 222-164-4
missing translation for 'rtecsNumber' : AJ1900000
missing translation for 'tsca' : TSCA
- Palladium(II) acetate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- LONG-TERM AQUATIC HAZARD Chronic 4
Hazard Statement
- H318-Causes serious eye damage.
- H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only