Learn More
Invitrogen™ Human PAPP-A ELISA Kit
ELISA
Brand: Invitrogen™ EHPAPPA
Description
The Human PAPP-A ELISA quantitates Hu PAPP-A in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu PAPP-A. Principle of the method The Human PAPP-A solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Pregnancy-associated plasma protein-A (PAPP-A, Insulin-like growth factor binding protein-4 protease, PAPP-A-proMBP complex) is a large heterottetrameric glycoprotein of approximately 500 kDa, which was first discovered in serum from pregnant women. The hetreotetramer consists of two PAPP-A subunits and two proMBP subunits. Low maternal serum levels of PAPP-A in first trimester biochemical screening is used as a marker of Down's syndrome (trisomy 21).
Specifications
Q13219 | |
0.6 ng/mL | |
Solid-phase sandwich ELISA | |
ELISA | |
Plasma, Serum, Supernatant | |
96 assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
5069 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
96 Tests | |
Differentiation | |
2°C to 8°C | |
PAPPA | |
4 hrs 45 mins |
0.686 to 500 ng/mL | |
0.6 ng/mL to 500 ng/mL | |
HRP | |
Other Proteins | |
This ELISA kit shows no cross-reactivity with the following cytokines tested: human Alpha-2 Microglobulin, ApoA, ApoC1, ApoC2, ApoC3, ApoE, ApoH, Artemin, BMP-6, Calbindin, CK beta 8-1 (CCL23), CNTF, EMMPRIN (CD147), Endostatin, Fibrinogen, FLRG, TNFSF18 (GITR Ligand), GPBB, Hemoglobin, HTRA2, IL-17 E, IL-1 R3 (IL1 R Acp), Lactoferrin, Mesothelin, PAPP-A, PD-ECGF (Thymidine Phosphorylase), PDGF R alpha, Pepsinogen 2, Periostin, Persephin, PTH, RAP, SAA (Serum Amyloid A), Serpin F1 (PEDF), Tie-1, TPA, Thrombospondin 2, Ubiquitin+1, Vitronectin, VWF | |
Human | |
Colorimetric Microplate Reader | |
ASBABP2,DIPLA1,IGFBP-4 ase,PAPA,PAPP-A1,PAPPA | |
<10 | |
HRP | |
RUO | |
Plasma 50 μL, Serum 50 μL, Supernatant 100 μL | |
RP11-45A16.1, ASBABP2, DIPLA1, IGFBP-4 ase, PAPA, PAPP-A1, PAPPA | |
1 hrs 20 mins |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.